Assay Details
| Gene Symbol: GATM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC039389.1 | - | 5 | 857 | 178 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928414 | LOC101928414 | -100.0kb | Anti-sense | Chr. 15 - 45251581 - 45279222 on NCBI Build GRCh38 | |
| Downstream | spermatogenesis associated 5 like 1 | SPATA5L1 | 22.7kb | Sense | Chr. 15 - 45400202 - 45421419 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 4631 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 52546 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 374747 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P115556 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P115556 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16787085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118715 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16787085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118715 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30118695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09626220 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115556 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115556 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P115556 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 92244 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 49011 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P115556 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P115556 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1566860_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000458245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010865.hg.1 | ENST00000527933.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1557692_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557692_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566860_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566860_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566861_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566861_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566860_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566861_at4 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052860 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052860 | - | - |

GATM
Genomic Map