Assay Details
| Gene Symbol: PECAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000442.4 | NP_000433.4 | 13 - 14 | 2324 | 67 | |
| XM_005276880.1 | XP_005276937.1 | 13 - 14 | 2610 | 67 | ||
| XM_011524889.2 | XP_011523191.1 | 14 - 15 | 2340 | 67 | ||
| XM_011524890.1 | XP_011523192.1 | 14 - 15 | 2289 | 67 | ||
| XM_017024738.1 | XP_016880227.1 | 14 - 15 | 2233 | 67 | ||
| XM_017024739.1 | XP_016880228.1 | 12 - 13 | 2556 | 67 | ||
| GenBank mRNA | AF281287.1 | - | 13 - 14 | 2194 | 67 | |
| AF281288.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281289.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281290.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281291.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281292.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281293.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281294.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281295.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281296.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281297.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281298.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281299.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281300.1 | - | 13 - 14 | 2194 | 67 | ||
| AF281301.1 | - | 13 - 14 | 2194 | 67 | ||
| AF393676.1 | - | 13 - 14 | 2194 | 67 | ||
| AF393677.1 | - | 13 - 14 | 2194 | 67 | ||
| AF393678.1 | - | 13 - 14 | 2194 | 67 | ||
| AK290692.1 | - | 13 - 14 | 2127 | 67 | ||
| AK303959.1 | - | 8 - 9 | 1098 | 67 | ||
| AK304166.1 | - | 12 - 13 | 2124 | 67 | ||
| BC022512.1 | - | 13 - 14 | 2304 | 4816878 | 67 | |
| BC051822.1 | - | 13 - 14 | 2324 | 5768579 | 67 | |
| JQ287500.1 | - | 13 - 14 | 2111 | 67 | ||
| M28526.1 | - | 13 - 14 | 2252 | 67 | ||
| M37780.1 | - | 13 - 14 | 2236 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PECAM1
Genomic Map