Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000309048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000462462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000464507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000602446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000602787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NM_001270517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NM_199294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NR_036462.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000309048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000462462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000464507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000602446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
ENST00000602787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NM_001270517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NM_199294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
NR_036462.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147289.hg.1 |
TC0100006819.hg.1 |
Clariom D TC |