Assay Details
Gene Symbol: APITD1-CORT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243768.1 | NP_001230697.1 | 3 - 4 | 754 | 98 | |
NM_198544.3 | NP_940946.1 | 4 - 5 | 690 | 98 | ||
GenBank mRNA | AF521016.1 | - | 4 - 5 | 690 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15022495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15022496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00041114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00041001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15022493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051604 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00041000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040989 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00041119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051497 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00040985 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20051601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00041115 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 58108 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 20117 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 307428 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 270270 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 576557 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 281066 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P121257 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P134446 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P121257 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P134446 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P30033384 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P00013131 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P258164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20051601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20051601 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P134446 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P121257 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040999 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00041002 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040996 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00041106 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040994 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040995 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040992 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00040997 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00013125 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P134446 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P121257 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00013121 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00013128 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P134446 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162598 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 31105 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00013125 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00013128 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00013130 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P134446 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041114 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041106 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041121 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041117 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041115 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041112 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041128 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041110 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041119 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041136 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041142 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041130 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041134 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041138 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041108 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00041125 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P134446 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00013125 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00013128 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_198544_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897645 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897647 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897638 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897636 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897634 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897646 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897633 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897637 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7897632 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 213454_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11743796_a_at | - | - |
Affymetrix Human Genome U219 Array | 11740303_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11743796_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11740303_a_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_198544_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015525 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015529 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015529 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Sentrix Human-6 Expression BeadChip | GI_41327699-S | NM_199006.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_41327699-S | AF516753.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_41327699-S | AF521016.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_41327699-S | NM_198544.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_41327699-S | NM_199294.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_10245-A | - | - |
Illumina human-6 v1.0 expression beadchip | 0000460050 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0000460050 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1793360 | NM_199006.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793360 | AF516753.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793360 | AF521016.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793360 | NM_198544.3 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793360 | CR593499.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793360 | NM_199006.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793360 | AF516753.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793360 | AF521016.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793360 | NM_198544.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793360 | CR593499.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2650372 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_10245 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793360 | NM_199006.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793360 | AF516753.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793360 | AF521016.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793360 | NM_198544.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793360 | CR593499.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793360 | NM_199006.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793360 | AF516753.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793360 | AF521016.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793360 | NM_198544.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793360 | CR593499.1 | human_transcriptome |
