Assay Details
| Gene Symbol: FYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199335.3 | NP_955367.1 | 12 - 13 | 2097 | 76 | |
| XM_011514011.2 | XP_011512313.1 | 12 - 13 | 2058 | 76 | ||
| XM_017009316.1 | XP_016864805.1 | 12 - 13 | 2057 | 76 | ||
| GenBank mRNA | AF001862.1 | - | 11 - 12 | 2001 | 76 | |
| BX647195.1 | - | 12 - 13 | 2082 | 76 | ||
| BX647196.1 | - | 12 - 13 | 2083 | 76 | ||
| U93049.1 | - | 11 - 12 | 1964 | 76 | ||
More Information
Set Membership: |
|

FYB
Genomic Map