Assay Details
| Gene Symbol: ABCG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529806.1 | XP_011528108.1 | 15 | 5945 | 62 | |
| XM_011529807.2 | XP_011528109.1 | 15 | 5967 | 62 | ||
| GenBank mRNA | AL355688.1 | - | 1 | 1122 | 62 | |
| AL355711.1 | - | 1 | 1122 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABCG1
Genomic Map