Assay Details
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011514074.2 | XP_011512376.1 | 2 - 3 | 928 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011514074.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
