Assay Details
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011514074.2 | XP_011512376.1 | 3 - 4 | 1318 | 105 | |
XM_011514075.2 | XP_011512377.1 | 3 - 4 | 1314 | 105 | ||
GenBank mRNA | BC013351.2 | - | 2 - 3 | 263 | 3621124 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC013351.2,XM_011514074.2,XM_011514075.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174306.hg.1 | TC0500010640.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174305.hg.1 | TC0500010640.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000397080.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000500337.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000506860.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | ENST00000512085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | NM_022483.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174304.hg.1 | TC0500010640.hg.1 | Clariom D TC |
