Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC013351.2 | - | 1 - 2 | 117 | 3621124 | 93 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC013351.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174309.hg.1 | TC0500010640.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500174308.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174308.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174308.hg.1 | TC0500010640.hg.1 | Clariom D TC |

TMEM267
Genomic Map