Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
TC07000868.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
ENST00000343526.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
ENST00000415680.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
ENST00000452999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
ENST00000536822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
NM_003852.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013512.hg.1 |
NM_015905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
TC07000868.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
ENST00000343526.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
ENST00000415680.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
ENST00000536822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
NM_003852.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013511.hg.1 |
NM_015905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000343526.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000415680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000620194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
NM_003852.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
NM_015905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000343526.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000415680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
ENST00000620194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
NM_003852.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
NM_015905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166177.hg.1 |
TC0700009317.hg.1 |
Clariom D TC |