Assay Details
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ278977.1 | - | 2 - 3 | 129 | ||
AJ430614.1 | - | 4 | 258 | |||
JX015383.1 | - | 3 | 169 | 141 | ||
JX015384.1 | - | 3 | 169 | 141 | ||
JX015385.1 | - | 3 | 250 | 222 | ||
JX015387.1 | - | 4 | 310 | 282 | ||
U33202.1 | - | 2 | 163 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
