Assay Details
| Gene Symbol: NOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000620.4 | NP_000611.1 | 24 - 25 | 4417 | 87 | |
| NM_001204213.1 | NP_001191142.1 | 23 - 24 | 3009 | 87 | ||
| NM_001204214.1 | NP_001191143.1 | 23 - 24 | 3004 | 87 | ||
| NM_001204218.1 | NP_001191147.1 | 25 - 26 | 4519 | 87 | ||
| XM_011538398.2 | XP_011536700.1 | 26 - 27 | 4390 | 87 | ||
| XM_017019345.1 | XP_016874834.1 | 25 - 26 | 4526 | 87 | ||
| XM_017019346.1 | XP_016874835.1 | 25 - 26 | 4340 | 87 | ||
| XM_017019347.1 | XP_016874836.1 | 24 - 25 | 4327 | 87 | ||
| GenBank mRNA | D16408.1 | - | 23 - 24 | 3706 | 87 | |
| EU753241.1 | - | 24 - 25 | 4391 | 87 | ||
| L02881.1 | - | 23 - 24 | 4133 | 87 | ||
| U17327.1 | - | 24 - 25 | 4391 | 87 | ||
| U31466.1 | - | 23 - 24 | 3713 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002014.hg.1
Set Membership: |
|

NOS1
Genomic Map