Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324255.1 | NP_001311184.1 | 4 - 5 | 7010 | 71 | |
| NM_005909.4 | NP_005900.2 | 5 - 6 | 7280 | 71 | ||
| GenBank mRNA | CR933720.1 | - | 1 - 2 | 4928 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000336.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule bundle formation
cellular process
dendrite development
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
establishment of monopolar cell polarity
structural molecule activity
protein binding
microtubule binding
microtubule bundle formation
cellular process
dendrite development
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
establishment of monopolar cell polarity
structural molecule activity
protein binding
microtubule binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05004598.hg.1 | TC05000336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004598.hg.1 | ENST00000296755.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004598.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152717.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152717.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152717.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152717.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152717.hg.1 | TC0500007738.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000296755.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000504492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000511641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000512974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | ENST00000513526.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007738.hg.1 | NM_005909.4 | Refseq |

MAP1B
Genomic Map