Assay Details
| Gene Symbol: SNRPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003091.3 | NP_003082.1 | 6 - 7 | 853 | 155 | |
| GenBank mRNA | BC080516.1 | - | 6 - 7 | 771 | 2822685 | 155 |
| J04564.1 | - | 6 - 7 | 806 | 148 | ||
| M34081.1 | - | 5 - 6 | 669 | 155 | ||
| X17567.1 | - | 6 - 7 | 744 | 155 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001744.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
positive regulation of defense response to virus by host
termination of RNA polymerase II transcription
protein methylation
histone mRNA metabolic process
RNA splicing
nuclear import
mitophagy in response to mitochondrial depolarization
xenophagy
protein binding
U2 snRNA binding
poly(A) RNA binding
telomerase RNA binding
histone pre-mRNA DCP binding
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
positive regulation of defense response to virus by host
termination of RNA polymerase II transcription
protein methylation
histone mRNA metabolic process
RNA splicing
nuclear import
mitophagy in response to mitochondrial depolarization
xenophagy
protein binding
U2 snRNA binding
poly(A) RNA binding
telomerase RNA binding
histone pre-mRNA DCP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | TC20001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | ENST00000303103.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | ENST00000381342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | ENST00000438552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | ENST00000474384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008090.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008093.hg.1 | TC20001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008093.hg.1 | ENST00000303103.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008093.hg.1 | ENST00000381342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008093.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | TC20001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | ENST00000303103.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | ENST00000339610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | ENST00000381342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | ENST00000438552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | ENST00000474384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008098.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011623.hg.1 | ENST00000443634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | TC2000009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170942.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | TC2000009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170940.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009972.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011623.hg.1 | ENST00000443634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | TC2000009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170938.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | NM_198216.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | TC2000009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000339610.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000381342.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000438552.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | ENST00000474384.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | NM_003091.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170939.hg.1 | NM_198216.1 | Refseq |

SNRPB
Genomic Map