Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144974.3 | NP_659411.2 | 6 - 7 | 185 | ||
| XM_017020397.1 | XP_016875886.1 | 6 - 7 | 185 | |||
| GenBank mRNA | AW373287.1 | - | 1 - 2 | 184 | 185 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13007023.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007023.hg.1 | TC13000614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000614023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007024.hg.1 | ENST00000444614.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007024.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007024.hg.1 | TC13000614.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300159145.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159145.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159145.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159145.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159145.hg.1 | TC1300008772.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300159146.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159146.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159146.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159146.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159146.hg.1 | TC1300008772.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000614023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 190096 | - | - |

CCDC122
Genomic Map