Assay Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025112.4 | NP_079388.3 | 8 - 9 | 2450 | 150 | |
| NR_104249.1 | 7 - 8 | 2630 | 150 | |||
| XM_011513119.2 | XP_011511421.1 | 7 - 8 | 2298 | 150 | ||
| GenBank mRNA | AK023923.1 | - | 5 - 6 | 1152 | 150 | |
| AY277596.1 | - | 4 - 5 | 1012 | 150 | ||
| AY936556.1 | - | 8 - 9 | 2410 | 150 | ||
| BC002940.2 | - | 7 - 8 | 1388 | 150 | ||
| BC012729.2 | - | 7 - 8 | 1388 | 150 | ||
| BC063713.1 | - | 7 - 8 | 1698 | 150 | ||
| DQ079590.1 | - | 8 - 9 | 2450 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001739.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03031615.hg.1 | TC03001739.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031615.hg.1 | ENST00000389709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031615.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000336332.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_001040653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012281.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000389709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000514463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | ENST00000515545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | NM_025112.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | NR_104249.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191870.hg.1 | TC0300012281.hg.1 | Clariom D TC |

ZXDC
Genomic Map