Assay Details
Gene Symbol: RNLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031709.2 | NP_001026879.2 | 1 - 2 | 252 | 85 | |
NM_018363.3 | NP_060833.1 | 1 - 2 | 252 | 85 | ||
XM_005269946.1 | XP_005270003.1 | 1 - 2 | 178 | 85 | ||
XM_005269947.1 | XP_005270004.1 | 1 - 2 | 178 | 85 | ||
XM_005269949.4 | XP_005270006.1 | 1 - 2 | 178 | 85 | ||
XM_011539924.2 | XP_011538226.1 | 1 - 2 | 178 | 85 | ||
XM_011539927.2 | XP_011538229.1 | 1 - 2 | 178 | 85 | ||
XM_017016380.1 | XP_016871869.1 | 1 - 2 | 178 | 85 | ||
XM_017016381.1 | XP_016871870.1 | 1 - 2 | 178 | 85 | ||
XM_017016383.1 | XP_016871872.1 | 1 - 2 | 178 | 85 | ||
GenBank mRNA | AK002080.1 | - | 1 - 2 | 179 | 85 | |
BC005364.1 | - | 1 - 2 | 3828014 | 85 | ||
BP332262.1 | - | 1 - 2 | 252 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001495.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | ENST00000331772.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | ENST00000371947.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | ENST00000437752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | ENST00000466945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018908.hg.1 | TC10001495.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018910.hg.1 | ENST00000371947.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018910.hg.1 | ENST00000466945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018910.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018910.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018910.hg.1 | TC10001495.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | ENST00000331772.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | ENST00000371947.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | ENST00000466945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10018907.hg.1 | TC10001495.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177204.hg.1 | TC1000011363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000331772.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000437752.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177205.hg.1 | TC1000011363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000371947.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000466945.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | ENST00000481793.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | NM_001031709.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | NM_018363.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177207.hg.1 | TC1000011363.hg.1 | Clariom D TC |
