Assay Details
| Gene Symbol: RNLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269948.2 | XP_005270005.1 | 1 - 2 | 789 | 70 | |
| XM_005269950.3 | XP_005270007.1 | 1 - 2 | 789 | 70 | ||
| XM_017016382.1 | XP_016871871.1 | 1 - 2 | 789 | 70 | ||
| XM_017016384.1 | XP_016871873.1 | 1 - 2 | 789 | 70 | ||
| GenBank mRNA | AK296262.1 | - | 1 - 2 | 187 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000331772.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000371947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000437752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000466945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | ENST00000481793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_001031709.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011363.hg.1 | NM_018363.3 | Refseq |

RNLS
Genomic Map