Assay Details
| Gene Symbol: STAMBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020799.3 | NP_065850.1 | 2 - 3 | 534 | 142 | |
| XM_006717928.2 | XP_006717991.1 | 3 - 4 | 635 | 142 | ||
| XM_017016457.1 | XP_016871946.1 | 2 - 3 | 881 | 142 | ||
| GenBank mRNA | AB037794.1 | - | 2 - 3 | 855 | 142 | |
| AK056086.1 | - | 2 - 3 | 463 | 142 | ||
| BC010846.1 | - | 2 - 3 | 452 | 3898533 | 142 | |
| CD244526.1 | - | 2 - 3 | 559 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056086.1, BC010846.1, NM_020799.3, XM_006717928.2, XM_017016457.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156807.hg.1 | TC1000008385.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156809.hg.1 | TC1000008385.hg.1 | Clariom D TC |

STAMBPL1
Genomic Map