Assay Details
| Gene Symbol: STAMBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB037794.1 | - | 10 - 11 | 2077 | 72 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |

STAMBPL1
Genomic Map