Assay Details
| Gene Symbol: ARHGAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004815.3 | NP_004806.3 | 12 - 13 | 1653 | 90 | |
| XM_006711048.3 | XP_006711111.1 | 11 - 12 | 1287 | 90 | ||
| XM_011542438.2 | XP_011540740.1 | 12 - 13 | 1528 | 90 | ||
| XM_011542439.2 | XP_011540741.1 | 12 - 13 | 1373 | 90 | ||
| XM_011542440.2 | XP_011540742.1 | 11 - 12 | 1247 | 90 | ||
| GenBank mRNA | AA326126.1 | - | 2 - 3 | 281 | 90 | |
| AB208873.1 | - | 11 - 12 | 1259 | 90 | ||
| AK307733.1 | - | 12 - 13 | 1482 | 90 | ||
| AK310491.1 | - | 12 - 13 | 1373 | 90 | ||
| BC093741.1 | - | 11 - 12 | 1326 | 7939586 | 90 | |
| BC093767.1 | - | 11 - 12 | 1329 | 7939612 | 90 | |
| BX508890.1 | - | 5 - 6 | 583 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002884.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045056.hg.1 | TC01002884.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045056.hg.1 | ENST00000260526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045056.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045056.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242356.hg.1 | TC0100018454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 107454 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 219040 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 232959 | - | - |

ARHGAP29
Genomic Map