Assay Details
| Gene Symbol: PPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021129.3 | NP_066952.1 | 10 - 11 | 945 | 127 | |
| GenBank mRNA | AB026723.1 | - | 10 - 11 | 868 | 127 | |
| AF092439.1 | - | 10 - 11 | 915 | 127 | ||
| AF119665.1 | - | 10 - 11 | 915 | 127 | ||
| AF154065.1 | - | 10 - 11 | 883 | 127 | ||
| AF217186.1 | - | 10 - 11 | 838 | 127 | ||
| AK058193.1 | - | 10 - 11 | 938 | 127 | ||
| BC001022.2 | - | 10 - 11 | 920 | 3345496 | 127 | |
| BC061581.1 | - | 10 - 11 | 928 | 6526628 | 127 | |
| BC105034.1 | - | 10 - 11 | 911 | 8144037 | 127 | |
| BC105036.1 | - | 10 - 11 | 911 | 8144039 | 127 | |
| BC107882.1 | - | 10 - 11 | 879 | 6495107 | 127 | |
| EU794626.1 | - | 10 - 11 | 945 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PPA1
Genomic Map