Assay Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020846.1 | NP_065897.1 | 2 - 3 | 678 | 128 | |
| XM_005248121.2 | XP_005248178.1 | 2 - 3 | 794 | 128 | ||
| GenBank mRNA | AB040891.1 | - | 2 - 3 | 648 | 128 | |
| AK026476.1 | - | 2 - 3 | 225 | 128 | ||
| AK291006.1 | - | 2 - 3 | 808 | 128 | ||
| BC040993.1 | - | 2 - 3 | 957 | 5272534 | 128 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000278.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04004005.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004005.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004005.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004005.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004005.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150867.hg.1 | TC0400007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |

SLAIN2
Genomic Map