Assay Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020846.1 | NP_065897.1 | 4 - 5 | 1007 | 57 | |
| XM_005248121.2 | XP_005248178.1 | 4 - 5 | 1123 | 57 | ||
| GenBank mRNA | AB040891.1 | - | 4 - 5 | 977 | 57 | |
| AK026476.1 | - | 4 - 5 | 554 | 57 | ||
| AK291006.1 | - | 4 - 5 | 1137 | 57 | ||
| BC031691.2 | - | 2 - 3 | 327 | 57 | ||
| BC040993.1 | - | 4 - 5 | 1286 | 5272534 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000278.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC031691.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04004011.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004011.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004011.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004011.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004010.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004010.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004010.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004010.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004012.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004012.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004012.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004012.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150875.hg.1 | TC0400007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150874.hg.1 | TC0400007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150873.hg.1 | TC0400007447.hg.1 | Clariom D TC |

SLAIN2
Genomic Map