Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022361.4 | NP_071756.2 | 1 - 2 | 147 | 76 | |
GenBank mRNA | AK314101.1 | - | 1 - 2 | 147 | 76 | |
BF244570.1 | - | 160 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001977.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314101.1,NM_022361.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06024527.hg.1 | TC06001977.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024527.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024527.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024527.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024522.hg.1 | TC06001977.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024522.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024522.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024528.hg.1 | TC06001977.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024528.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024528.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024523.hg.1 | TC06001977.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024523.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024523.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024523.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024523.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187671.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187671.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187671.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187671.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187671.hg.1 | TC0600012709.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000429112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012709.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187664.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187664.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187664.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187664.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187664.hg.1 | TC0600012709.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187670.hg.1 | TC0600012709.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000254765.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000474760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | ENST00000489134.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | NM_022361.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | NR_024539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187666.hg.1 | TC0600012709.hg.1 | Clariom D TC |
