Assay Details
| Gene Symbol: BVES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199563.1 | NP_001186492.1 | 6 - 7 | 1035 | 72 | |
| NM_007073.4 | NP_009004.2 | 6 - 7 | 905 | 72 | ||
| NM_147147.3 | NP_671488.1 | 6 - 7 | 975 | 72 | ||
| XM_011535398.2 | XP_011533700.1 | 6 - 7 | 910 | 72 | ||
| GenBank mRNA | AF204172.1 | - | 6 - 7 | 929 | 72 | |
| AK289979.1 | - | 6 - 7 | 1019 | 72 | ||
| BC034425.1 | - | 6 - 7 | 975 | 4821891 | 72 | |
| BC040502.1 | - | 6 - 7 | 1035 | 5295907 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001976.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor recycling
regulation of heart rate
hematopoietic progenitor cell differentiation
response to ischemia
heart development
muscle organ development
skeletal muscle tissue development
regulation of cell shape
vesicle-mediated transport
substrate adhesion-dependent cell spreading
positive regulation of locomotion
regulation of membrane potential
regulation of GTPase activity
vesicle docking
sinoatrial node cell development
cell migration involved in heart development
epithelial cell-cell adhesion
regulation of endocytic recycling
structural molecule activity
protein binding
cAMP binding
positive regulation of receptor recycling
regulation of heart rate
hematopoietic progenitor cell differentiation
response to ischemia
heart development
muscle organ development
skeletal muscle tissue development
regulation of cell shape
vesicle-mediated transport
substrate adhesion-dependent cell spreading
positive regulation of locomotion
regulation of membrane potential
regulation of GTPase activity
vesicle docking
sinoatrial node cell development
cell migration involved in heart development
epithelial cell-cell adhesion
regulation of endocytic recycling
structural molecule activity
protein binding
cAMP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | TC06001976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | ENST00000314641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | ENST00000336775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024503.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012708.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000314641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000336775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | ENST00000446408.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_001199563.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_007073.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | NM_147147.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187644.hg.1 | TC0600012708.hg.1 | Clariom D TC |

BVES
Genomic Map