Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006800.hg.1 |
ENST00000432905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006800.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006800.hg.1 |
TC13000596.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006798.hg.1 |
ENST00000432905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006798.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006798.hg.1 |
TC13000596.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
ENST00000432905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
ENST00000458118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
ENST00000432905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
ENST00000458118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158719.hg.1 |
TC1300008697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009950.hg.1 |
ENST00000445572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
ENST00000432905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
ENST00000458118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
ENST00000432905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
ENST00000458118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
NR_038258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158723.hg.1 |
TC1300008697.hg.1 |
Clariom D TC |