Assay Details
Gene Symbol: LINC00858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038220.1 | 2 - 3 | 286 | 90 | ||
GenBank mRNA | AK056561.1 | - | 2 - 3 | 286 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000595.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10006513.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006513.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006513.hg.1 | TC10000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10006512.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006512.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006512.hg.1 | TC10000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006511.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006511.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006511.hg.1 | TC10000595.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | ENST00000504333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010838.hg.1 | NR_109904.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156081.hg.1 | TC1000008274.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156082.hg.1 | TC1000008274.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156083.hg.1 | TC1000008274.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
