Assay Details
| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038220.1 | 5 - 6 | 1268 | 133 | ||
| GenBank mRNA | AK056561.1 | - | 5 - 6 | 1268 | 133 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000595.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362366 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362323 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362391 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16366152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16366154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16366151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362339 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16366150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07362378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27749538 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 75138 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 83951 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 314508 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 697458 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3718352 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37464 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3718352 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3718352 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P19002887 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19002887 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139973 | AJ005371.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139973 | AK314980.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139973 | AY306129.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139973 | BC036373.2 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139973 | NM_014364.4 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 29015 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19002887 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39112752 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19002874 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19002887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563821_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006518.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006518.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006518.hg.1 | TC10000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156088.hg.1 | TC1000008274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000415469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | ENST00000437892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008274.hg.1 | NR_038220.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1563821_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563821_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563821_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044211 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 712335 | - | - |

LINC00858
Genomic Map