Assay Details
| Gene Symbol: SLCO3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011521456.2 | XP_011519758.1 | 2 - 3 | 186 | 82 | |
| GenBank mRNA | AK097797.1 | - | 2 - 3 | 186 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK097797.1, XM_011521456.2 transcripts.
Set Membership: |
|

SLCO3A1
Genomic Map