Assay Details
| Gene Symbol: FAM9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171186.1 | NP_001164657.1 | 8 - 9 | 1060 | 121 | |
| NM_174951.3 | NP_777611.1 | 8 - 9 | 1035 | 121 | ||
| GenBank mRNA | AF494343.1 | - | 8 - 9 | 1035 | 121 | |
| BC112022.1 | - | 8 - 9 | 1001 | 8327490 | 121 | |
| BC113449.1 | - | 8 - 9 | 989 | 8322501 | 121 | |
| BC143805.1 | - | 8 - 9 | 1060 | 9052320 | 121 | |
More Information
Set Membership: |
|

FAM9A
Genomic Map