Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020399.1 | XP_016875888.1 | 6 - 7 | 1052 | 167 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | ENST00000614023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000444614.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000470137.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000476570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000611737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | ENST00000614023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008772.hg.1 | NM_144974.4 | Refseq |

CCDC122
Genomic Map