Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168687.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168687.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168687.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168687.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168687.hg.1 |
TC0200009832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009832.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168685.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168685.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168685.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168685.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168685.hg.1 |
TC0200009832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200168686.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168686.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168686.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168686.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168686.hg.1 |
TC0200009832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200168684.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168684.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168684.hg.1 |
ENST00000446838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200168684.hg.1 |
NR_110258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200168684.hg.1 |
TC0200009832.hg.1 |
Clariom D TC |