Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020397.1 | XP_016875886.1 | 1 - 2 | 177 | 88 | |
| XM_017020398.1 | XP_016875887.1 | 1 - 2 | 177 | 88 | ||
| XM_017020399.1 | XP_016875888.1 | 1 - 2 | 177 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017020397.1, XM_017020398.1, XM_017020399.1 transcripts.
Set Membership: |
|

CCDC122
Genomic Map