Assay Details
| Gene Symbol: ESCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052911.2 | NP_443143.2 | 1 - 2 | 103 | 137 | |
| XM_011525799.2 | XP_011524101.1 | 1 - 2 | 366 | 137 | ||
| GenBank mRNA | AB067498.1 | - | 1 - 2 | 39 | 137 | |
| AK096746.1 | - | 1 - 2 | 107 | 136 | ||
| AL832041.1 | - | 1 - 2 | 256 | 137 | ||
| BG492713.1 | - | 1 - 2 | 126 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000401.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB067498.1,AL832041.1,NM_052911.2,XM_011525799.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004114.hg.1 | ENST00000269214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004114.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004114.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004114.hg.1 | TC18000401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004115.hg.1 | ENST00000269214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004115.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004115.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004115.hg.1 | TC18000401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | TC1800008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156219.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | TC1800008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156216.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | TC1800008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156220.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |

ESCO1
Genomic Map