Assay Details
Gene Symbol: ESCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052911.2 | NP_443143.2 | 3 - 4 | 350 | 132 | |
XM_011525798.1 | XP_011524100.1 | 3 - 4 | 480 | 132 | ||
XM_011525799.2 | XP_011524101.1 | 3 - 4 | 613 | 132 | ||
GenBank mRNA | AB067498.1 | - | 3 - 4 | 286 | 132 | |
AK096746.1 | - | 3 - 4 | 353 | 132 | ||
AL832041.1 | - | 3 - 4 | 503 | 132 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000401.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB067498.1,AL832041.1,NM_052911.2,XM_011525798.1,XM_011525799.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18004113.hg.1 | ENST00000269214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004113.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004113.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004113.hg.1 | TC18000401.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004112.hg.1 | ENST00000269214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004112.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004112.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004112.hg.1 | TC18000401.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18004111.hg.1 | ENST00000269214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004111.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18004111.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18004111.hg.1 | TC18000401.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | TC1800008231.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156212.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | TC1800008231.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156213.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | TC1800008231.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156214.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | NM_052911.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | TC1800008231.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | ENST00000269214.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | ENST00000383276.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156215.hg.1 | NM_052911.2 | Refseq |
