Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
ENST00000372801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
ENST00000372806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
ENST00000396731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
ENST00000499879.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004878.hg.1 |
TC20000339.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
TC2000007476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151954.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000372806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000474717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000480745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000487587.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000488618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
ENST00000499879.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007476.hg.1 |
NM_006282.4 |
Refseq |