Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
TC04000510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000264572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000339360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000380158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000408900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000408927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
ENST00000453712.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_001100426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_001100427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_001100428.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_001100429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_001100430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007495.hg.1 |
NM_021159.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000264572.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000339360.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000380158.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000408900.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000408927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000453712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100427.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100428.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_021159.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000264572.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000339360.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000380158.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000408900.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000408927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
ENST00000453712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100427.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100428.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_001100430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
NM_021159.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156505.hg.1 |
TC0400008213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000264572.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000339360.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000380158.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000408900.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000408927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000453712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000509501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100427.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100428.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_021159.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000264572.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000339360.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000380158.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000408900.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000408927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000453712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
ENST00000509501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100427.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100428.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_001100430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
NM_021159.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156504.hg.1 |
TC0400008213.hg.1 |
Clariom D TC |