Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000067.2 | NP_000058.1 | 3 - 4 | 529 | 79 | |
GenBank mRNA | AK312978.1 | - | 3 - 4 | 424 | 79 | |
BC011949.1 | - | 3 - 4 | 414 | 3863603 | 79 | |
BC035424.1 | - | 2 - 3 | 319 | 79 | ||
CR536526.1 | - | 3 - 4 | 349 | 79 | ||
CR541875.1 | - | 3 - 4 | 349 | 79 | ||
FJ224289.1 | - | 3 - 4 | 529 | 79 | ||
GQ472235.1 | - | 3 - 4 | 414 | 79 | ||
J03037.1 | - | 3 - 4 | 414 | 79 | ||
M36532.1 | - | 3 - 4 | 393 | 79 | ||
Y00339.1 | - | 3 - 4 | 387 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000531.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
morphogenesis of an epithelium
one-carbon metabolic process
response to pH
response to zinc ion
carbon dioxide transport
bicarbonate transport
positive regulation of synaptic transmission, GABAergic
positive regulation of cellular pH reduction
angiotensin-activated signaling pathway
odontogenesis of dentin-containing tooth
response to estrogen
regulation of anion transport
positive regulation of osteoclast differentiation
positive regulation of bone resorption
secretion
response to steroid hormone
regulation of intracellular pH
cellular response to fluid shear stress
positive regulation of dipeptide transmembrane transport
regulation of chloride transport
arylesterase activity
carbonate dehydratase activity
protein binding
zinc ion binding
kidney development
morphogenesis of an epithelium
one-carbon metabolic process
response to pH
response to zinc ion
carbon dioxide transport
bicarbonate transport
positive regulation of synaptic transmission, GABAergic
positive regulation of cellular pH reduction
angiotensin-activated signaling pathway
odontogenesis of dentin-containing tooth
response to estrogen
regulation of anion transport
positive regulation of osteoclast differentiation
positive regulation of bone resorption
secretion
response to steroid hormone
regulation of intracellular pH
cellular response to fluid shear stress
positive regulation of dipeptide transmembrane transport
regulation of chloride transport
arylesterase activity
carbonate dehydratase activity
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08007209.hg.1 | TC08000531.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08007209.hg.1 | ENST00000285379.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007209.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000518231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008126.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000285379.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000520127.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000520996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | ENST00000522742.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | NM_000067.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | NM_001293675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155789.hg.1 | TC0800008126.hg.1 | Clariom D TC |
