| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002296.hg.1 |
TC04002929.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002296.hg.1 |
ENST00000265016.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002296.hg.1 |
ENST00000382346.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002296.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012769.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000265016.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000382346.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000505785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000514445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
ENST00000514989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
NM_004334.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187927.hg.1 |
TC0400012769.hg.1 |
Clariom D TC |