Assay Details
Gene Symbol: SLC26A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000441.1 | NP_000432.1 | 6 - 7 | 986 | 151 | |
XM_005250425.2 | XP_005250482.1 | 6 - 7 | 969 | 151 | ||
XM_006716025.3 | XP_006716088.1 | 5 - 6 | 765 | 151 | ||
XM_017012318.1 | XP_016867807.1 | 5 - 6 | 765 | 151 | ||
GenBank mRNA | AF030880.1 | - | 6 - 7 | 986 | 151 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000695.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
regulation of pH
sensory perception of sound
sulfate transport
inorganic anion transport
bicarbonate transport
iodide transport
oxalate transport
regulation of protein localization
regulation of membrane potential
regulation of intracellular pH
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
iodide transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
ion transport
regulation of pH
sensory perception of sound
sulfate transport
inorganic anion transport
bicarbonate transport
iodide transport
oxalate transport
regulation of protein localization
regulation of membrane potential
regulation of intracellular pH
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
iodide transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07010895.hg.1 | TC07000695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07010895.hg.1 | ENST00000265715.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07010895.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07010896.hg.1 | TC07000695.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07010896.hg.1 | ENST00000265715.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07010896.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161921.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161921.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161921.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161921.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161921.hg.1 | TC0700008760.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700161920.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161920.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161920.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161920.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161920.hg.1 | TC0700008760.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000265715.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000440056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000460748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000477350.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000480841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000492030.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | ENST00000497446.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008760.hg.1 | NM_000441.1 | Refseq |
