Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003009.hg.1 |
NM_012157.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000538892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003010.hg.1 |
NM_012157.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000507198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000538892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
NM_001171713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003007.hg.1 |
NM_012157.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003008.hg.1 |
NM_012157.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000507198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000538892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
NM_001171713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003011.hg.1 |
NM_012157.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
TC03000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
ENST00000283627.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
ENST00000446237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
ENST00000484457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
ENST00000538181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
ENST00000542085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003006.hg.1 |
NM_012157.4 |
Refseq |