Assay Details
| Gene Symbol: IL13RA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000640.2 | NP_000631.1 | 1 - 2 | 85 | 91 | |
| GenBank mRNA | AK291957.1 | - | 1 - 2 | 54 | 91 | |
| AK309444.1 | - | 1 - 2 | 95 | 91 | ||
| U70981.1 | - | 1 - 2 | 53 | 91 | ||
| Y08768.1 | - | 1 - 2 | 85 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001290.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291957.1,NM_000640.2,U70981.1,Y08768.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017449.hg.1 | TC0X001290.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017449.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017449.hg.1 | ENST00000371936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017449.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017449.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017452.hg.1 | TC0X001290.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017452.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017452.hg.1 | ENST00000371936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017452.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017452.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010557.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171921.hg.1 | TC0X00010557.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171917.hg.1 | TC0X00010557.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000243213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000371936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | ENST00000468224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | NM_000640.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171918.hg.1 | TC0X00010557.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199239 | - | - |

IL13RA2
Genomic Map