Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000411471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000422643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000486511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000611578.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000614832.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000616380.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000619012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NM_001280795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NM_004445.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NR_104001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000411471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000422643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000486511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000611578.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000614832.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000616380.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
ENST00000619012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NM_001280795.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NM_004445.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
NR_104001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167279.hg.1 |
TC0700009472.hg.1 |
Clariom D TC |