Assay Details
Gene Symbol: LRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018214.4 | NP_060684.4 | 2 - 3 | 555 | 78 | |
XM_011514726.2 | XP_011513028.1 | 3 - 4 | 919 | 78 | ||
XM_017010997.1 | XP_016866486.1 | 2 - 3 | 847 | 78 | ||
GenBank mRNA | AF332199.1 | - | 2 - 3 | 502 | 78 | |
AF359380.1 | - | 2 - 3 | 278 | 78 | ||
AK001637.1 | - | 2 - 3 | 259 | 78 | ||
AK021896.1 | - | 3 - 4 | 919 | 78 | ||
AK292248.1 | - | 2 - 3 | 400 | 78 | ||
BC003193.1 | - | 2 - 3 | 555 | 3504090 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000668.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007991.hg.1 | TC06000668.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007991.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007991.hg.1 | ENST00000370888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007991.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157947.hg.1 | TC0600008303.hg.1 | Clariom D TC |
