Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007997.hg.1 |
TC06000668.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007997.hg.1 |
ENST00000370888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007997.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157953.hg.1 |
TC0600008303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000370888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000487251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
ENST00000490222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008303.hg.1 |
NM_018214.4 |
Refseq |