Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
ENST00000290291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
ENST00000357163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
ENST00000370461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
ENST00000370468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
ENST00000370469.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006992.hg.1 |
TC20000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
ENST00000290291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
ENST00000357163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
ENST00000370461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
ENST00000370468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
ENST00000370469.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006995.hg.1 |
TC20000498.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000450048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008030.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
NM_007346.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
TC2000008030.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000290291.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000370461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
ENST00000621591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155911.hg.1 |
NM_007346.3 |
Refseq |