| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006828.hg.1 |
TC08000499.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006828.hg.1 |
ENST00000354370.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006828.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006828.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155105.hg.1 |
TC0800008002.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000354370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396419.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000396423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
ENST00000494318.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008002.hg.1 |
NM_004133.4 |
Refseq |