Assay Details
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005065.5 | NP_005056.3 | 15 - 16 | 1607 | 97 | |
XM_005267988.2 | XP_005268045.1 | 15 - 16 | 1490 | 97 | ||
XM_005267989.3 | XP_005268046.1 | 14 - 15 | 1541 | 97 | ||
GenBank mRNA | AB020335.1 | - | 15 - 16 | 1535 | 97 | |
AF052059.1 | - | 15 - 16 | 1583 | 97 | ||
AK075511.1 | - | 15 - 16 | 1586 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001372.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14015552.hg.1 | ENST00000336735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14015552.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14015552.hg.1 | TC14001372.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400172661.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400172661.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400172661.hg.1 | ENST00000336735.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400172661.hg.1 | NM_005065.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400172661.hg.1 | TC1400009842.hg.1 | Clariom D TC |
