Assay Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005065.5 | NP_005056.3 | 18 - 19 | 1984 | 66 | |
| XM_005267988.2 | XP_005268045.1 | 18 - 19 | 1867 | 66 | ||
| XM_005267989.3 | XP_005268046.1 | 17 - 18 | 1918 | 66 | ||
| GenBank mRNA | AB020335.1 | - | 18 - 19 | 1912 | 66 | |
| AF052059.1 | - | 18 - 19 | 1960 | 66 | ||
| BC040498.1 | - | 1 - 2 | 71 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001372.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015548.hg.1 | ENST00000261258.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015548.hg.1 | ENST00000336735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015548.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015548.hg.1 | TC14001372.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015549.hg.1 | ENST00000336735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015549.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015549.hg.1 | TC14001372.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172657.hg.1 | TC1400009842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |

SEL1L
Genomic Map